missing translation for 'onlineSavingsMsg'
Learn More

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%, Thermo Scientific Chemicals

Catalog Number 15595126
Click to view available options
Quantity:
100 mg

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 510758-28-8
Melting Point 138°C to 140°C
Color Brown
Assay Percent Range 96% min. (HPLC)
Molecular Formula C30H30N10
MDL Number MFCD09265124
Quantity 100 mg
Synonym tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine, 1,2,4-trimethyl-5-benzenesulfonic acid, tris 1-benzyl-1h-1,2,3-triazole-4-ylmethyl amine, tris 1-benzyl-1h-1,2,3-triazol-4-yl methyl amine tbta
InChI Key WKGZJBVXZWCZQC-UHFFFAOYSA-N
SMILES C(N(CC1=CN(CC2=CC=CC=C2)N=N1)CC1=CN(CC2=CC=CC=C2)N=N1)C1=CN(CC2=CC=CC=C2)N=N1
IUPAC Name 1-(1-benzyltriazol-4-yl)-N,N-bis[(1-benzyltriazol-4-yl)methyl]methanamine
Molecular Weight (g/mol) 530.64
PubChem CID 11203363
Formula Weight 530.64
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Show More Show Less
compliance-icons
Product Identifier
  • Tris¢(1-benzyl-1H-1,2,3-triazol-4-yl)methyl!amine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'