missing translation for 'onlineSavingsMsg'
Learn More

Triruthenium dodecacarbonyl, 99%, Thermo Scientific Chemicals

Catalog Number p-3737521
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15243-33-1
C12O12Ru3
639.33
MFCD00011209
NQZFAUXPNWSLBI-UHFFFAOYSA-N
Ruthenium carbonyl
cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1
This item is not returnable. View return policy
15243-33-1
C12O12Ru3
639.33
MFCD00011209
NQZFAUXPNWSLBI-UHFFFAOYSA-N
Ruthenium carbonyl
cyclotriruthenium; dodecakis(methanidylidyneoxidanium)
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Ru]1[Ru][Ru]1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 150.0°C
Color Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Ru3(CO)12
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 639.33
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Triruthenium dodecacarbonyl
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'