missing translation for 'onlineSavingsMsg'
Learn More

Tripropyl orthoformate, 98%, Thermo Scientific™

Catalog Number 10622364
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
621-76-1
C10H22O3
190.29
MFCD00015214
RWNXXQFJBALKAX-UHFFFAOYSA-N
tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
69311
1-(dipropoxymethoxy)propane
CCCOC(OCCC)OCCC
This item is not returnable. View return policy
621-76-1
C10H22O3
190.29
MFCD00015214
RWNXXQFJBALKAX-UHFFFAOYSA-N
tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
69311
1-(dipropoxymethoxy)propane
CCCOC(OCCC)OCCC
Alkylation reagent
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8790g/mL
Boiling Point 106°C to 108°C (40.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH(OCH2CH2CH3)3
Refractive Index 1.4060 to 1.408
Quantity 100 mL
Beilstein 02, IV, 27
Specific Gravity 0.879
Formula Weight 190.29
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tripropyl orthoformate
Show More Show Less
compliance-icons
Product Identifier
  • Tripropyl orthoformate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'