missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphosphopyridine nucleotide, disodium salt, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% min. (enzymatic,on dry subst.) |
Packaging | Glass bottle |
Quantity | 100 mg |
Solubility Information | (0.05% in water) Clear colorless to light yellow |
IUPAC Name | disodium 1-[(2R,3R,4S,5R)-5-({[({[(2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-(hydrogen phosphonatooxy)-3-hydroxyoxolan-2-yl]methyl phosphonato}oxy)(oxido)phosphoryl]oxy}methyl)-3,4-dihydroxyoxolan-2-yl]-3-carbamoyl-1λ⁵-pyridin-1-ylium |
Formula Weight | 787.4 |
Percent Purity | 97+% |
Physical Form | Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'