missing translation for 'onlineSavingsMsg'
Learn More

Triphenylphosphinecopper(I) hydride hexamer, 96%, Thermo Scientific Chemicals

Catalog Number p-3213307
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33636-93-0
C108H96Cu6P6
1961.07
MFCD00221518
IZZFKTUYPRVQID-UHFFFAOYSA-T
stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
11982471
copper;triphenylphosphane
[H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
33636-93-0
C108H96Cu6P6
1961.07
MFCD00221518
IZZFKTUYPRVQID-UHFFFAOYSA-T
stryker's reagent, triphenylphosphine copper hydride hexamer, triphenylphosphine-copper i hydride hexamer, unii-8x3r32m7xu, hydrido triphenylphosphine copper i hexamer, stryker's reagent mi, triphenylphosphine cuprous hydride, triphenylphosphine copper i hydride hexamer, copper, hexa-mu3-hydrohexakis triphenylphosphine hexa-, octahedro, cuprous hydride triphenylphosphine hexamer
11982471
copper;triphenylphosphane
[H+].[H+].[H+].[H+].[H+].[H+].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)[Cu]123[Cu]45([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]11([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]22([P+](C6=CC=CC=C6)(C6=CC=CC=C6)C6=CC=CC=C6)[Cu]34([P+](C3=CC=CC=C3)(C3=CC=CC=C3)C3=CC=CC=C3)[Cu]512[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Color Red
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass Bottle
Quantity 1 g
Merck Index 15, 8987
Solubility Information Solubility in water: reacts slowly.
Formula Weight 1961.07
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material Triphenylphosphinecopper(I) hydride hexamer
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'