missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Triphenylphosphine diiodide, tech. 90%

Brand:  Alfa Aesar™ H55585.06

Code : NEW

Additional Details : CAS Number : 6396-07-2 Weight : 0.00500kg Transport : UN number : 2923 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15471888

  • £75.20 / 5g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Triphenylphosphine diiodide
Technical Grade
516.1
UN2923
Diiodotriphenylphosphorane
6396-07-2
(C6H5)3PI2
MFCD00216568
2740417

Specifications

Triphenylphosphine diiodide
Technical Grade
90%
516.1
UN2923
Diiodotriphenylphosphorane
210° to 211°C
Light Sensitive
6396-07-2
Powder
(C6H5)3PI2
MFCD00216568
2740417
Yellow to orange
5g
Videos
SDS