missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylphosphine dibromide, ca. 33% bromine, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Melting Point | 235.0°C |
Color | Pink to Yellow |
Infrared Spectrum | Authentic |
Packaging | Plastic bottle |
Linear Formula | (C6H5)3PBr2 |
Quantity | 100 g |
Beilstein | 16, 864 |
Fieser | 01,1247; 13,333 |
Merck Index | 15, 9917 |
Formula Weight | 422.09 |
Show More |
Safety and Handling
Product Identifier
- Triphenylphosphine dibromide, ca. 33% bromine
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'