Learn More
Triphenylphosphine, 99+%, Thermo Scientific Chemicals
Description
Triphenylphosphine is used in the synthesis of organic compounds due to its nucleophilicity and its reducing character. It is involved in the synthesis of biaryl compounds, phosphonium salts and other phosphorus compounds. As a reducing agent, it is used to prepare aromatic amines from the corresponding aromatic N-oxides. The anionic phosphine is usually isolated as the trisodium salt which reacts with rhodium to form a complex that finds use in industrial hydroformylation reactions. It is also used to prepare Wilkinson's catalyst, RhCl(PPh3)3 useful to catalyze the hydrogenation of alkenes and tetrakis(triphenylphosphine)palladium(0) that is widely used to catalyze C-C coupling reactions in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 79°C to 81°C |
Density | 1.2 g/mL |
Boiling Point | 360°C |
Flash Point | 181°C (358°F) |
Assay Percent Range | >99% |
Quantity | 25 g |
Beilstein | 610776 |
Solubility Information | Soluble in ether,benzene,carbon tetrachloride,glacial acetic acid,acetone,chloroform and alcohol. Insoluble in water. |
Formula Weight | 262.29 |
Physical Form | Flakes |
Show More |
RUO – Research Use Only