missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenylphosphine, 99%, Thermo Scientific Chemicals
365.25 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.
Use promo code "23251" to get your promotional price.
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 78.5°C to 81.5°C |
Color | White |
Boiling Point | 377.0°C |
Flash Point | 181°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Plastic drum |
Linear Formula | (C6H5)3P |
Quantity | 5 kg |
Beilstein | 16,759 |
Show More |
Safety and Handling
Product Identifier
- Triphenylphosphine
- Warning
- Acute toxicity Category 4
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H317-May cause an allergic skin reaction.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'