missing translation for 'onlineSavingsMsg'
Learn More

Triphenylmethane, 98%, Thermo Scientific Chemicals

Catalog Number p-7081653
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
519-73-3
C19H16
244.34
MFCD00004763
AAAQKTZKLRYKHR-UHFFFAOYSA-N
triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
10614
CHEBI:76212
benzhydrylbenzene
C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
519-73-3
C19H16
244.34
MFCD00004763
AAAQKTZKLRYKHR-UHFFFAOYSA-N
triphenylmethane, tritane, benzene, 1,1',1-methylidynetris, methane, triphenyl, diphenylmethyl benzene, 1,1',1-methanetriyltribenzene, unii-8o4utw9e17, ccris 5194, triphenyl methane, triphenylmethane?
10614
CHEBI:76212
benzhydrylbenzene
C1=CC=C(C=C1)C(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 92°C to 94°C
Density 1.01
Boiling Point 358°C to 359°C
Quantity 25 g
Beilstein 1909753
Merck Index 14,9741
Formula Weight 244.34
Percent Purity 98%
Chemical Name or Material Triphenylmethane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'