missing translation for 'onlineSavingsMsg'
Learn More

Triphenylborane, 96%, Thermo Scientific Chemicals

Catalog Number p-7049984
Click to view available options
Quantity:
1 g
5 g
25 g
960-71-4
C18H15B
242.128
MFCD00003007
MXSVLWZRHLXFKH-UHFFFAOYSA-N
triphenylboron, borane, triphenyl, triphenyboron, triphenylborine, borine, triphenyl 6ci, boron triphenyl, triphenyl boron, triphenyl-boran, borine,triphenyl, triphenyl-borane
70400
triphenylborane
B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3
960-71-4
C18H15B
242.128
MFCD00003007
MXSVLWZRHLXFKH-UHFFFAOYSA-N
triphenylboron, borane, triphenyl, triphenyboron, triphenylborine, borine, triphenyl 6ci, boron triphenyl, triphenyl boron, triphenyl-boran, borine,triphenyl, triphenyl-borane
70400
triphenylborane
B(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Triphenylborane is used as a precursor in the production of pyridine-triphenylborane complex, which is used as a catalyst for the polymerization of acrylic esters. It is also used in the hydrocyanation of butadiene to adiponitrile. It is involved in the synthesis of 3-phenyl-cyclohexene by reacting with acetic acid cyclohex-2-enyl ester in presence of palladium bis(dibenzylideneacetone) and triphenylphosphine as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 147°C
Boiling Point 208°C (14 mmHg)
Odor Odorless
Quantity 1 g
UN Number UN1325
Beilstein 1961313
Sensitivity Air Sensitive
Solubility Information Soluble in aromatic solvents. Insoluble in water.
Formula Weight 242.13
Percent Purity 96%
Physical Form Crystalline
Chemical Name or Material Triphenylborane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'