Learn More
Triphenylborane, 96%, Thermo Scientific Chemicals
Description
Triphenylborane is used as a precursor in the production of pyridine-triphenylborane complex, which is used as a catalyst for the polymerization of acrylic esters. It is also used in the hydrocyanation of butadiene to adiponitrile. It is involved in the synthesis of 3-phenyl-cyclohexene by reacting with acetic acid cyclohex-2-enyl ester in presence of palladium bis(dibenzylideneacetone) and triphenylphosphine as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 147°C |
Boiling Point | 208°C (14 mmHg) |
Odor | Odorless |
Quantity | 25 g |
UN Number | UN1325 |
Beilstein | 1961313 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in aromatic solvents. Insoluble in water. |
Formula Weight | 242.13 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only