missing translation for 'onlineSavingsMsg'
Learn More

Triphenylacetic Acid 99%, ACROS Organics™

Brand:  Acros Organics 140300050

Code : 29

Additional Details : CAS Number : 595-91-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10477340

  • £33.10 / 5g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

Triphenylacetic acid
595-91-5
99%
(C6H5)3CCO2H
MFCD00004185
99%
≥98.5%
C20H16O2
288.34

Specifications

Triphenylacetic acid
267° to 275°C
99%
Authentic
Fine Crystalline Powder
99%
(C6H5)3CCO2H
MFCD00004185
White to beige
5g
595-91-5
Glass bottle
≥98.5%
C20H16O2
288.34
09, 712
Videos
SDS