missing translation for 'onlineSavingsMsg'
Learn More

Triphenylacetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-208660
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
595-91-5
C20H16O2
288.35
MFCD00004185
DCYGAPKNVCQNOE-UHFFFAOYSA-N
triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
68992
2,2,2-triphenylacetic acid
OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
595-91-5
C20H16O2
288.35
MFCD00004185
DCYGAPKNVCQNOE-UHFFFAOYSA-N
triphenylacetic acid, acetic acid, triphenyl, triphenyl acetic acid, benzeneacetic acid, .alpha.,.alpha.-diphenyl, nsc 61, tri phenylacetic acid, pubchem9200, alpha-toluic acid, alpha,alpha-diphenyl, acmc-1arvb, maybridge1_006927
68992
2,2,2-triphenylacetic acid
OC(=O)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 268°C to 269°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C6H5)3CCO2H
Quantity 5 g
Beilstein 09, 712
Formula Weight 288.34
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Triphenylacetic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'