missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triphenyl phosphate, 98%, Thermo Scientific Chemicals
17.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1000 g
Description
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.
Solubility
Reacts with water.
Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Triphenyl phosphate is used as an internal standard in the screening and quantification of agrochemical residues in vegetables and fruits. It is used in plastic processing and molding of plastic flow and performance.
Solubility
Reacts with water.
Notes
Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
Specifications
Melting Point | 48°C to 52°C |
Boiling Point | 247°C to 250°C (11 mmHg) |
Flash Point | 223°C (433°F) |
Odor | Odorless |
Quantity | 250 g |
UN Number | UN3077 |
Beilstein | 1888236 |
Merck Index | 14,9742 |
Solubility Information | Reacts with water. |
Formula Weight | 326.29 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'