missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsulfoxonium iodide, 98%, Thermo Scientific Chemicals
33.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 175.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic bottle |
Linear Formula | (CH3)3S(O)I |
Quantity | 100 g |
Solubility Information | Solubility in water: 15g/L (20°C). Other solubilities: insoluble in ethanol and ether |
Formula Weight | 220.07 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Trimethylsulfoxonium iodide
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'