missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl trifluoroacetate, 96%, ACROS Organics™

Brand:  Acros Organics 215971000

Code : NEW

Additional Details : CAS Number : 400-53-3 Weight : 0.10000kg Transport : UN number : 1993 Chem class : 1 Pack group : II


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 15274684

  • £87.10 / 100g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket


Specifications

Chemical Identifiers

Trimethylsilyl trifluoroacetate
400-53-3
96%
CF3CO2Si(CH3)3
MFCD00000413
96%
96%
C5H9F3O2Si
186.21

Specifications

Trimethylsilyl trifluoroacetate
0°C
88.0° to 90.0°C
100g
400-53-3
Glass bottle
Powder
96%
CF3CO2Si(CH3)3
MFCD00000413
1.0800g/mL
1.08
White
96%
Authentic
1.3350 to 1.3380
96%
C5H9F3O2Si
186.21
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Trimethylsilyl trifluoroacetate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.