missing translation for 'onlineSavingsMsg'
Learn More

Trimethylsilyl isothiocyanate, 94%, Thermo Scientific Chemicals

Catalog Number p-7043880
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2290-65-5
C4H9NSSi
131.268
MFCD00004797
XLTUPERVRFLGLJ-UHFFFAOYSA-N
trimethylsilyl isothiocyanate, silane, isothiocyanatotrimethyl, trimethylsilylisothiocyanate, isothiocyanato trimethyl silane, tmsisothiocyanate, tms-isothiocyanate, acmc-1clhj, trimethylsilyl-isothiocyanate, trimethylsilyl isothio-cyanate
75297
isothiocyanato(trimethyl)silane
C[Si](C)(C)N=C=S
This item is not returnable. View return policy
2290-65-5
C4H9NSSi
131.268
MFCD00004797
XLTUPERVRFLGLJ-UHFFFAOYSA-N
trimethylsilyl isothiocyanate, silane, isothiocyanatotrimethyl, trimethylsilylisothiocyanate, isothiocyanato trimethyl silane, tmsisothiocyanate, tms-isothiocyanate, acmc-1clhj, trimethylsilyl-isothiocyanate, trimethylsilyl isothio-cyanate
75297
isothiocyanato(trimethyl)silane
C[Si](C)(C)N=C=S

Trimethylsilyl isothiocyanate is used as derivatizing reagent during the synthesis of amino acid thiohydantoins. It participates in ring opening reaction of N-substituted aziridines and cyclohexene oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylsilyl isothiocyanate is used as derivatizing reagent during the synthesis of amino acid thiohydantoins. It participates in ring opening reaction of N-substituted aziridines and cyclohexene oxide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Strong oxidizing agentsAlcohols, Strong bases, Amines, acids.
TRUSTED_SUSTAINABILITY
Melting Point -33°C
Density 0.932
Boiling Point 143°C to 144°C
Flash Point 29°C (84°F)
Odor Pungent
Refractive Index 1.482
Quantity 5 g
UN Number UN1992
Beilstein 1745262
Sensitivity Moisture sensitive; May form a precipitate
Solubility Information Reacts with water.
Formula Weight 131.27
Percent Purity 94%
Chemical Name or Material Trimethylsilyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'