missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylsilyl cyanide, 98%, Thermo Scientific Chemicals
712.03 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Boiling Point | 114.0°C to 117.0°C |
Melting Point | 11.0°C |
Quantity | 500 g |
Assay Percent Range | 98% |
Linear Formula | (CH3)3SiCN |
Fieser | 04,542; 05,720; 06,632; 07,397; 08,133; 09,127; 10,112; 11,147; 12,148; 13,87; 14,107; 15,102; 17,89; 01,979; 04,418; 06,503; 08,433 |
Flash Point | 1°C |
Solubility Information | Solubility in water: reacts. |
Formula Weight | 99.21 |
Percent Purity | 98% |
Show More |
Safety and Handling
Product Identifier
- Trimethylsilyl cyanide
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 2
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH029-Contact with water liberates toxic gas.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'