Trimethylsilyl acetate 97%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 391980250

25GR Trimethylsilyl acetate, 97%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 2754-27-0 Weight: 0.07500kg Transport: UN number : 3272 Chem class : 1 Pack group : II


Packaging Glass bottle
Quantity 25g

Product Code. 10004973

Quantity Price
1 £ 14.5 / 25g
Estimated Shipment date
from Supplier 23-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Trimethylsilyl acetate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H12O2Si
Linear Formula CH3COOSi(CH3)3
Formula Weight 132.23
MDL Number MFCD00008695

Specification

Assay ≥96.0%
Physical Form Liquid
Formula Weight 132.23
Quantity 25g
Color Clear colorless to pale yellow
Linear Formula CH3COOSi(CH3)3
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 2754-27-0
Infrared Spectrum Authentic
Refractive Index 1.3870 to 1.3890 (20°C, 589nm)
Assay Percent Range 97%
MDL Number MFCD00008695
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Highly flammable liquid and vapor.
Molecular Formula C5H12O2Si
Health Hazard 1 GHS Signal Word: Danger
Density 0.884g/mL
Chemical Name or Material Trimethylsilyl acetate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.