missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylgermanium chloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Trimethylgermanium chloride is used to prepare trimethyl(2-thienyl)germane. It is also used as alkylating and precursor to prepare Grignard reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -14°C |
Density | 1.249 g/mL |
Boiling Point | 102°C |
Flash Point | 1°C (35°F) |
Refractive Index | 1.4337 |
Quantity | 5 g |
UN Number | UN2924 |
Sensitivity | Moisture sensitive |
Solubility Information | Immiscible with water.Miscible with ether,tetrahydrofuran,hexane,benzene,dichloromethane and chloroform. |
Formula Weight | 153.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'