missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl orthoformate, 99+%, anhydrous, AcroSeal™, Thermo Scientific Chemicals
17.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
800 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -53.0°C |
Density | 0.9700g/mL |
Boiling Point | 101.0°C to 102.0°C |
Flash Point | 13°C |
Assay Percent Range | 99% min. (GC) |
Packaging | AcroSeal glass Bottle |
Linear Formula | CH(OCH3)3 |
Refractive Index | 1.3780 to 1.3800 |
Quantity | 100 mL |
Merck Index | 15,698 |
Show More |
Safety and Handling
Product Identifier
- Trimethyl orthoformate
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'