missing translation for 'onlineSavingsMsg'
Learn More

Trimethylboroxine, 3.5M (50 wt.%) solution in THF, Thermo Scientific Chemicals

Catalog Number p-206986
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
823-96-1, 109-99-9
C3H9B3O3
125.53
MFCD00013354
GBBSAMQTQCPOBF-UHFFFAOYSA-N
trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
574072
2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
CB1OB(C)OB(C)O1
This item is not returnable. View return policy
823-96-1, 109-99-9
C3H9B3O3
125.53
MFCD00013354
GBBSAMQTQCPOBF-UHFFFAOYSA-N
trimethylboroxine, 2,4,6-trimethylboroxin, trimethylboroxin, boroxin, trimethyl, trimethyl-1,3,5,2,4,6-trioxatriborinane, trimethyboroxine, trimethyl boroxin, trimethylboroxaine, trimethyl boroxine, trimethyl-boroxine
574072
2,4,6-trimethyl-1,3,5,2,4,6-trioxatriborinane
CB1OB(C)OB(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -38.0°C
Color Colorless to Yellow
Density 0.8900g/mL
Boiling Point 78.0°C to 80.0°C
Flash Point −9°C
Packaging Glass bottle
Quantity 25 mL
Specific Gravity 0.89
Formula Weight 125.54
Percent Purity 40.0 to 60.0% (ex Boron)
Physical Form Solution
Chemical Name or Material Trimethylboroxine, 50 wt. % Solution in THF
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'