missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethylbenzylammoniumhexachloroplatinate(IV), Premion™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 043474.03

Code : NEW

Additional Details : CAS Number : 41909-89-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11395787

  • £111.41 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Trimethylbenzylammoniumhexachloroplatinate (IV)
Premion™
708.28
41909-89-1
[C6H5CH2N(CH3)3]2PtCl6
MFCD00054403

Specifications

Trimethylbenzylammoniumhexachloroplatinate (IV)
41909-89-1
Powder
(metals basis)
708.28
1g
Premion™
99.99%
[C6H5CH2N(CH3)3]2PtCl6
MFCD00054403
Videos
SDS