missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethylbenzylammoniumhexachloroplatinate(IV), Premion™, 99.99% (metals basis)

Brand:  Alfa Aesar™ 043474.06

Code : NEW

Additional Details : CAS Number : 41909-89-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11305797

  • £489.40 / 5g


Estimated Shipment date
from Supplier 07-06-2018
Add to basket

Specifications

Chemical Identifiers

Trimethylbenzylammoniumhexachloroplatinate (IV)
Premion™
708.28
41909-89-1
[C6H5CH2N(CH3)3]2PtCl6
MFCD00054403

Specifications

Trimethylbenzylammoniumhexachloroplatinate (IV)
41909-89-1
Powder
(metals basis)
708.28
5g
Premion™
99.99%
[C6H5CH2N(CH3)3]2PtCl6
MFCD00054403
Videos
SDS