missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, ACROS Organics™

Brand:  Acros Organics 270440050

Code : 29

Additional Details : CAS Number : 75-50-3 Weight : 3.80000kg Transport : UN number : 2924 Chem class : 1b Pack group : II

Quantity 5L

 View more versions of this product

Product Code. 10733154

  • £300.50 / 5 L


Estimated Shipment
16-03-2018
Add to basket

Specifications

Chemical Identifiers

Trimethylamine
Pure
C3H9N
59.11
64-17-5
32 to 34%
(CH3)3N
MFCD00008327

Specifications

Trimethylamine
−20°C
Clear colorless
64-17-5
Pure
32 to 34%
(CH3)3N
MFCD00008327
0.7500g/mL
50.0°C
5L
1.3630 to 1.3660
Liquid
C3H9N
59.11
Videos
SDS