missing translation for 'onlineSavingsMsg'
Learn More

Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, ACROS Organics™

Brand:  Acros Organics 270440250

Code : 29

Additional Details : CAS Number : 75-50-3 Weight : 0.06875kg Transport : UN number : 2924 Chem class : 3 Pack group : II

Quantity 25mL

 View more versions of this product

Product Code. 10450521

  • £12.10 / 25mL


EU Stock 13
Add to basket

Specifications

Chemical Identifiers

Trimethylamine
Pure
C3H9N
59.11
64-17-5
32 to 34%
(CH3)3N
MFCD00008327

Specifications

Trimethylamine
−20°C
Clear colorless
64-17-5
1.3630 to 1.3660
Liquid
C3H9N
59.11
0.7500g/mL
50.0°C
25mL
Glass bottle
Pure
32 to 34%
(CH3)3N
MFCD00008327
Videos
SDS