missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals

Catalog Number 10522744
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
3282-30-2
C5H9ClO
120.58
MFCD00000709
JVSFQJZRHXAUGT-UHFFFAOYSA-N
pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride
62493
2,2-dimethylpropanoyl chloride
CC(C)(C)C(Cl)=O
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
3282-30-2
C5H9ClO
120.58
MFCD00000709
JVSFQJZRHXAUGT-UHFFFAOYSA-N
pivaloyl chloride, trimethylacetyl chloride, propanoyl chloride, 2,2-dimethyl, pivalyl chloride, pivaloylchloride, 2,2-dimethylpropionyl chloride, pivalic acid chloride, pivalolyl chloride, unii-jq82j0o21t, 2,2-dimethylpropionylchloride
62493
2,2-dimethylpropanoyl chloride
CC(C)(C)C(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -56.0°C
Color Colorless to Pink
Density 0.9800g/mL
Boiling Point 105.0°C
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCOCl
Refractive Index 1.4110 to 1.4130
Quantity 1 kg
Beilstein 02, 320
Fieser 01,1229; 08,404; 11,489
Specific Gravity 0.98
Solubility Information Solubility in water: hydrolysis., miscible with most common aprotic organic solvents
Viscosity 0.86 mPa.s (20°C)
Formula Weight 120.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Trimethylacetyl chloride, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Trimethylacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'