missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetonitrile, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081844
27.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
630-18-2
C5H9N
83.134
MFCD00001847
JAMNHZBIQDNHMM-UHFFFAOYSA-N
pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
12416
2,2-dimethylpropanenitrile
CC(C)(C)C#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
630-18-2
C5H9N
83.134
MFCD00001847
JAMNHZBIQDNHMM-UHFFFAOYSA-N
pivalonitrile, trimethylacetonitrile, 2,2-dimethylpropionitrile, tert-butyl cyanide, propanenitrile, 2,2-dimethyl, tert-butylnitrile, 2-cyano-2-methylpropane, trimethyl acetonitrile, tert-cyanobutane, t-bucn
12416
2,2-dimethylpropanenitrile
CC(C)(C)C#N

Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethylacetonitrile used as a solvent and as a labile ligand in coordination chemistry. It is also used as an intermediate in organic synthesis. It is used in the presence of titanium(II) chloride and zinc to undergoes reductive coupling reaction of aromatic and aliphatic ketones including acyclic aliphatic ketones to give the corresponding pinacols.

Solubility
Miscible with ethanol, acetone and toluene. Slightly miscible with water.

Notes
Store in a cool place. Incompatible with strong acids, strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 16°C to 18°C
Density 0.757
Boiling Point 105°C to 106°C
Flash Point 4°C (39°F)
Odor Odorless
Quantity 25 g
UN Number UN3273
Solubility Information Miscible with ethanol,acetone and toluene. Slightly miscible with water.
Formula Weight 83.13
Percent Purity ≥98%
Chemical Name or Material Trimethylacetonitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'