missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethyl phosphite, 97%

Brand:  Alfa Aesar™ 030132.36

Code : NEW

Additional Details : CAS Number : 121-45-9 Weight : 0.50000kg Transport : UN number : 2329 Chem class : 1 Pack group : III

Quantity 500g

 View more versions of this product

Product Code. 11332878

  • £38.48 / 500g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

Trimethyl phosphite
C3H9O3P
124.08
UN2329
121-45-9
(CH3O)3P
MFCD00008350
Methyl phosphite

Specifications

Trimethyl phosphite
27° (80°F)
111° to 112°C
−78°C
121-45-9
1.4080
97%
C3H9O3P
124.08
UN2329
1.052g/mL
Air & Moisture Sensitive
Colorless
500g
Unpleasant
Liquid
97%
(CH3O)3P
MFCD00008350
Methyl phosphite
Videos
SDS