missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Trimethyl borate, 70% azeotrope with methanol

Brand:  Alfa Aesar™ 044721.K7

Code : NEW

Additional Details : CAS Number : 121-43-7 Weight : 0.01000kg Transport : UN number : 1993 Chem class : 3 Pack group : II

Quantity 4L

 View more versions of this product

Product Code. 11409950

  • £90.10 / 4 L


Estimated Shipment date
from Supplier 19-06-2018
Add to basket

Specifications

Chemical Identifiers

Trimethyl borate
(CH3O)3B
MFCD00008346
14,9712
121-43-7
103.92
UN1993

Specifications

Trimethyl borate
0.885g/mL
Characteristic
4L
Liquid
103.92
UN1993
70% azeotrope with methanol
−9°C (16°F)
52° to 54°C
121-43-7
(CH3O)3B
MFCD00008346
14,9712
Videos
SDS