Learn More
TRIM56 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555061
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PECRETVPVP PEGVASFKTN FFVNGLLDLV KARACGDLRA GKPACALCPL VGGTSTGGPA TARCLDCADD LCQACADGHR C Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 29%.
TRIM56 Polyclonal antibody specifically detects TRIM56 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TRIM56 | |
Polyclonal | |
Unconjugated | |
TRIM56 | |
E3 ubiquitin-protein ligase TRIM56; RING finger protein 109; RING-type E3 ubiquitin transferase TRIM56; RNF109; TRIM56; tripartite motif containing 56; tripartite motif-containing protein 56 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81844 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRZ2 | |
TRIM56 | |
Recombinant protein corresponding to Human TRIM56 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only