missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl phosphite, 96%, ACROS Organics™

Brand:  Acros Organics 372380050

Code : 29

Additional Details : CAS Number : 116-17-6 Weight : 0.06000kg Transport : UN number : 3278 Chem class : 1 Pack group : III

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10221153

  • £16.50 / 5g


EU Stock 6
Add to basket

Specifications

Chemical Identifiers

Triisopropyl phosphite
96%
[(CH3)2CHO]3P
96%
C9H21O3P
208.24

Specifications

Triisopropyl phosphite
62°C
5g
Authentic
1.4095 to 1.4115
96%
C9H21O3P
208.24
0.9140g/mL
Clear
116-17-6
Glass bottle
Liquid
96%
[(CH3)2CHO]3P
Solubility in water: insoluble.
Videos
SDS