missing translation for 'onlineSavingsMsg'
Learn More

Triiron dodecacarbonyl, 99%, stabilized, ACROS Organics™

Brand:  Acros Organics 209290250

Code : 29

Additional Details : CAS Number : 17685-52-8 Weight : 0.07500kg Transport : UN number : 3178 Chem class : 4.1 Pack group : II


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10416373

  • £145.50 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Triiron dodecacarbonyl
C12Fe3O12
503.66
99%
Fe3(CO)12
MFCD00135617

Specifications

Triiron dodecacarbonyl
Black
25g
Authentic
Glass bottle
Powder or Crystals
99%
Fe3(CO)12
MFCD00135617
1.9900g/mL
140.0°C
17685-52-8
98.5% min.
5 to 10% MeOH
99%
C12Fe3O12
503.66
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Triiron dodecacarbonyl

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
  • Specific target organ toxicity Category 2

Hazard Statement
  • H371-May cause damage to organs.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS