missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluorothymidine, 98%, Thermo Scientific Chemicals
85.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 190°C to 193°C |
Assay Percent Range | 97.5% min. (HPLC) |
Quantity | 250 mg |
Merck Index | 159855 |
Solubility Information | Solubility in water: soluble. Other solubilities: slightly soluble in isopropanol |
Formula Weight | 296.2 |
Percent Purity | 98% |
Chemical Name or Material | Trifluorothymidine |
Safety and Handling
Product Identifier
- Trifluorothymidine
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'