Alfa Aesar™ Trifluoroiodomethane, 99.9%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 039665.30

250GR Trifluoroiodomethane, 99.9% 250g

UNSPSC: 12352101

Code: NEW

Additional Details:
Additional Details: CAS Number: 2314-97-8 Weight: 0.25000kg


Quantity 250g

Product Code. 15451547

Quantity Price
1 £ 928.56 / 250g
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Trifluoroiodomethane
CAS 2314-97-8
Formula Weight 195.91
MDL Number MFCD00001060
UN Number UN1956
Beilstein 1732740
Synonym Iodotrifluoromethane; Trifluoromethyl iodide

Specification

Formula Weight 195.91
Health Hazard 3 GHS P Statement
P281-P201-P308+P313-P405-P410+P403-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Protect from sunlight.
Store in a well-ventilated place.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.3790
Beilstein 1732740
Chemical Name or Material Trifluoroiodomethane
TSCA TSCA
Packaging Cylinder
Physical Form Gas
Assay 99.9%
MDL Number MFCD00001060
CAS 2314-97-8
UN Number UN1956
Synonym Iodotrifluoromethane; Trifluoromethyl iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.
DOT Information Transport Hazard Class: 2; Proper Shipping Name: COMPRESSED GAS N.O.S.
RTECS Number PB6975000
Density 2.36g/mL
Sensitivity Light Sensitive
Boiling Point −22.5°C
Quantity 250g
Safety and Handling

Warning

GHS H Statement
H280-H341
Contains gas under pressure
may explode if heated.
Suspected of causing genetic defects.

GHS P Statement
P281-P201-P308+P313-P405-P410+P403-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Protect from sunlight.
Store in a well-ventilated place.
Dispose of contents/container in accordance with local/regional/national/international regulations.