missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog Number p-4259088
17.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
250 g
500 g
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
407-25-0
C4F6O3
210.03
MFCD00000416
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
9845
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
FC(F)(F)C(=O)OC(=O)C(F)(F)F
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
407-25-0
C4F6O3
210.03
MFCD00000416
QAEDZJGFFMLHHQ-UHFFFAOYSA-N
trifluoroacetic anhydride, trifluoroacetic acid anhydride, 2,2,2-trifluoroacetic anhydride, bis trifluoroacetic anhydride, acetic acid, trifluoro-, anhydride, perfluoroacetic anhydride, trifluoroacetyl anhydride, tfaa, hexafluoroacetic anhydride, anhydrid kyseliny trifluoroctove
9845
(2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate
FC(F)(F)C(=O)OC(=O)C(F)(F)F

Trifluoroacetic anhydride, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass Bottle, Boiling Point: 39.1 deg.C, Undesignated, Melting Point: -63.5 deg.C, Molecular Weight: 210.03, Percent Purity: 99+%, Assay Percent Range: 99% min. (GC), Beilstein: 02, II, 186, CAS: 407-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY
Melting Point -63.5°C
Density 1.5100g/mL
Boiling Point 39.1°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula (CF3CO)2O
Refractive Index 1.3
Quantity 25 g
Beilstein 02, II, 186
Fieser 01,1221; 03,308; 05,701; 06,616; 07,389; 09,484; 12,530; 14,151; 15,14; 01,373
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with polar organic solvents, partically soluble in unpolar hydrocarbons
Formula Weight 210.03
Vapor Pressure 141mbar at 20°C
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Trifluoroacetic anhydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic anhydride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P273-Avoid release to the environment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'