missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
10 mL
50 mL
Packaging:
Amber Glass
Ampule
Ampules
Description
Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL
- Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
- Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
- Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
- TFA is a common mobile phase additive that can function as an ion pair reagent
Specifications
Absorbance | 0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm |
pH | 0.45 |
Melting Point | -15°C |
Boiling Point | 72°C |
Color | Colorless |
Assay Percent Range | ≥99.5 % |
Linear Formula | CF3COOH |
Merck Index | 15, 9847 |
Formula Weight | 114.02 |
Percent Purity | ≥99.5% |
Show More |
Safety and Handling
Product Identifier
- TRIFLUOROACETIC ACID
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'