missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid-d, for NMR, packaged in 1 ml ampoules, 99.5 atom % D, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Ampule
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
Specifications
Physical Form | Liquid |
Color | Colorless |
Density | 1.5000g/mL |
Deuterium (D) | 99.4% min. |
Packaging | Ampule |
Water | 0.05% max. (Coulometric) |
Quantity | 10 x 1 mL |
CAS | 599-00-8 |
Molecular Formula | C2HF3O2 |
Linear Formula | CF3CO2D |
Show More |
Safety and Handling
Product Identifier
- Trifluoroacetic acid-d
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H332-Harmful if inhaled.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'