missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid-d, 99.5%(Isotopic), Thermo Scientific Chemicals
30.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
Description
Trifluoroacetic acid-d is employed as solvent to investigate the conformations of cyclic peptides by proton NMR spectra. It is also used to compose NMR solvent mixture to investigate the NMR spectra of high molecular weight polyesters such as polyglycolide (PG) and poly(ethylene terephthalate)and as a biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Density | 1.50 g/mL |
Odor | Acrid |
Packaging | Prescored Ampule |
Sensitivity | Hygroscopic |
Quantity | 10 g |
Melting Point | -15°C |
Boiling Point | 72°C |
UN Number | UN2699 |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'