missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™
66.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
42.60 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
66.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
42.60 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
66.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
25 mL
100 mL
Description
C2HF3O2, CAS Number-76-05-1, acetic acid, trifluoro, cf3cooh, kyselina trifluoroctova, perfluoroacetic acid, trifluoroacetic acid, trifluoroaceticacid, trifluoro acetic acid, trifluoro-acetic acid, trifluoroacetic acid, trifluoroethanoic acid, 100mL, 72 deg.C, CHEBI:45892, Colorless, 114.02g/mol
Specifications
pH | 0.45 |
Melting Point | -15°C |
Boiling Point | 72°C |
Color | Colorless |
IUPAC Name | trifluoroacetic acid |
Formula Weight | 114.02g/mol |
Quantity | 25 mL |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
Vapor Density | 3.9 |
Show More |
Safety and Handling
Product Identifier
- Trifluoroacetic acid
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'