missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetaldehyde methyl hemiacetal, 90%, technical, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.3600g/mL |
Boiling Point | 104.0°C to 106.0°C |
Flash Point | 42°C |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CF3CH(OCH3)OH |
Quantity | 5 g |
Specific Gravity | 1.36 |
Formula Weight | 130.07 |
Percent Purity | 90% |
Show More |
Safety and Handling
Product Identifier
- Trifluoroacetaldehyde methyl hemiacetal
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'