missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenol, 99%, Thermo Scientific™

Catalog Number p-3758872
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
444-30-4
C7H5F3O
162.11
MFCD00002222
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
67958
2-(trifluoromethyl)phenol
C1=CC=C(C(=C1)C(F)(F)F)O
This item is not returnable. View return policy
444-30-4
C7H5F3O
162.11
MFCD00002222
ZOQOPXVJANRGJZ-UHFFFAOYSA-N
2-trifluoromethyl phenol, 2-hydroxybenzotrifluoride, o-trifluoromethylphenol, o-hydroxybenzotrifluoride, alpha,alpha,alpha-trifluoro-o-cresol, phenol, 2-trifluoromethyl, o-trifluoromethoxy phenol, a,a,a-trifluoro-o-cresol, 2-hydroxy-alpha,alpha,alpha-trifluorotoluene
67958
2-(trifluoromethyl)phenol
C1=CC=C(C(=C1)C(F)(F)F)O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 45°C to 46°C
Color White
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CF3C6H4OH
Quantity 250 mg
Beilstein 06, III, 1263
Formula Weight 162.11
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-(Trifluoromethyl)phenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoromethyl)phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'