missing translation for 'onlineSavingsMsg'
Learn More

α,α,α-Trifluoro-m-cresol, 98+%, Thermo Scientific Chemicals

Catalog Number p-204314
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
98-17-9
C7H5F3O
162.11
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F
This item is not returnable. View return policy
98-17-9
C7H5F3O
162.11
MFCD00002299
UGEJOEBBMPOJMT-UHFFFAOYSA-N
3-trifluoromethyl phenol, 3-hydroxybenzotrifluoride, m-trifluoromethylphenol, m-hydroxybenzotrifluoride, phenol, 3-trifluoromethyl, m-trifluoromethyl phenol, alpha,alpha,alpha-trifluoro-m-cresol, a,a,a-trifluoro-m-cresol
7376
3-(trifluoromethyl)phenol
C1=CC(=CC(=C1)O)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -1.8°C
Color Yellow
Density 1.3300g/mL
Boiling Point 178°C to 179°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CF3C6H4OH
Refractive Index 1.4570 to 1.459
Quantity 25 mL
Beilstein 06, I, 187
Specific Gravity 1.33
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 162.11
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material α, α, α-Trifluoro-m-cresol, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • a,a,a-Trifluoro-m-cresol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'