missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Triethyltin bromide

Brand:  Alfa Aesar™ 071135.06

Code : NEW

Additional Details : CAS Number : 2767-54-6 Weight : 0.00500kg Transport : UN number : 2788 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11394819

  • £65.20 / 5g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Triethyltin bromide
(C2H5)3SnBr
MFCD00000052
Bromotriethylstannane
2767-54-6
285.79
UN2788

Specifications

Triethyltin bromide
−13.5°C
5g
1.63g/mL
Moisture Sensitive
(C2H5)3SnBr
MFCD00000052
Bromotriethylstannane
223° to 225°C
2767-54-6
99°C (210°F)
1.520
Liquid
285.79
UN2788
Videos
SDS