missing translation for 'onlineSavingsMsg'
Learn More

(Triethylsilyl)acetylene 97%, ACROS Organics™

Brand:  Acros Organics 368730010

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 1777-03-3 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 1 Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 10747403

  • £20.40 / 1g


Estimated Shipment
21-06-2017
Add to basket


Specifications Description & Specifications

Chemical Identifiers

(Triethylsilyl)acetylene
C8H16Si
MFCD00075373
97%
140.3
Ethynyltriethylsilane

Specifications

(Triethylsilyl)acetylene
1g
97%
140.3
Ethynyltriethylsilane
0.783g/cm3
1777-03-3
C8H16Si
MFCD00075373
Documentation

Technical Documents