missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylsilane, 99%, Thermo Scientific Chemicals
288.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Boiling Point | 107.0°C to 108.0°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 500 g |
Assay Percent Range | 99% |
Linear Formula | (C2H5)3SiH |
Beilstein | 04,625 |
Fieser | 01,1218; 02,433; 03,304; 04,530; 05,694; 12,482; 13,340; 14,322; 15,338; 16,356; 17,367 |
Flash Point | −3°C |
Merck Index | 15,9841 |
Show More |
Safety and Handling
Hazard Category
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'