missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyloxonium tetrafluoroborate, 95%, stab. with 3-5% diethyl ether, Thermo Scientific Chemicals
30.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of μ -amino esters from lactams.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.
Solubility
Soluble in dichloromethane.
Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Triethyloxonium tetrafluoroborate is used as a powerful alkylating agent, especially for ethylation. It is used for the modification of carboxyl residues in proteins. It is involved in the preparation of µ -amino esters from lactams.
Solubility
Soluble in dichloromethane.
Notes
Moisture sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | 84°C to 89°C |
Assay Percent Range | 95%, Stabilized with 3-5% diethyl ether |
Linear Formula | (CH3CH2)3OBF4 |
Quantity | 25 g |
UN Number | UN2925 |
Beilstein | 3598090 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dichloromethane. |
Formula Weight | 189.99 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'