missing translation for 'onlineSavingsMsg'
Learn More

Triethylgermanium hydride, 97%, Thermo Scientific Chemicals

Catalog Number 11304868
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1188-14-3
C6H16Ge
160.82
MFCD00077989
LTLGQZPDSQYFSB-UHFFFAOYSA-N
hydride, triethylgermyl, triethyl germanium hydride
6327150
triethylgermanium
CC[GeH](CC)CC
This item is not returnable. View return policy
1188-14-3
C6H16Ge
160.82
MFCD00077989
LTLGQZPDSQYFSB-UHFFFAOYSA-N
hydride, triethylgermyl, triethyl germanium hydride
6327150
triethylgermanium
CC[GeH](CC)CC

Co-reactant for the preparation of germanium acrylates

Co-reactant for the preparation of germanium acrylates. They are the Synthetic Reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.004 g/mL
Boiling Point 122°C
Flash Point 5°C (41°F)
Refractive Index 1.4382
Quantity 5 g
UN Number UN3399
Solubility Information Soluble in water( 46g/100 mL (26°C).
Formula Weight 160.78
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethylgermanium hydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'