missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 1,1,2-ethanetricarboxylate, 99%, Thermo Scientific Chemicals

Catalog Number 10636714
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7459-46-3
MFCD00009154
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
81961
triethyl ethane-1,1,2-tricarboxylate
CCOC(=O)CC(C(=O)OCC)C(=O)OCC
This item is not returnable. View return policy
7459-46-3
MFCD00009154
TVWZLLYAJDSSCJ-UHFFFAOYSA-N
triethyl 1,1,2-ethanetricarboxylate, 1,1,2-ethanetricarboxylic acid, triethyl ester, 2-ethoxycarbonyl-succinic acid diethyl ester, triethyl ethane tricarboxylate, 1,1,2-triethyl ethane-1,1,2-tricarboxylate, triethyl ethane-1,2,2-tricarboxylate, ethane-1,1,2-tricarboxylic acid triethyl ester, ethane-1,1,2-tricarboxylic acid, triethyl ester, diethyl 2-ethoxycarbonyl butane-1,4-dioate, acmc-209ovd
81961
triethyl ethane-1,1,2-tricarboxylate
CCOC(=O)CC(C(=O)OCC)C(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.0700g/mL
Boiling Point 99°C (0.5 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H18O6
Linear Formula C2H5O2CCH2CH(CO2C2H5)2
Refractive Index 1.4280 to 1.43
Quantity 5 g
Beilstein 02, 813
Specific Gravity 1.07
Molecular Weight (g/mol) 246.26
Formula Weight 246.26
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethyl 1,1,2-ethanetricarboxylate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'